Learn More
Medchemexpress LLC 3,5-dichloro-N-[[1-[[(4S)-2,2-dimethyloxan-4-yl]methyl]-4-fluoropiperidin-4-yl]methyl]benzamide | 1072018-68-8 | MFCD00078701 | 99.6% | 431.37 | C21H29Cl2FN2O2 | 1 ML
SDP
Catalog No. 5000214846
Encompass_Preferred
TTA-P2 is a selective, orally active, and brain-penetrant T-type calcium channel blocker used in neuroscience research to investigate neuronal excitability, pain mechanisms, and absence epilepsy. It is supplied as a 10 mM solution in DMSO or as solid material for assay preparation and in vivo studies.
  • Selective T-type calcium channel inhibitor with low-nanomolar potency (IC50 ≈ 22 nM).
  • Brain-penetrant profile suitable for in vivo studies.
  • Available as a 10 mM solution in DMSO and as solid quantities for flexible dosing.
  • High purity appropriate for research applications.
  • Used in studies of pain, tremor, and epilepsy mechanisms.
Catalog No. 50-002-14846 Supplier Medchemexpress LLC Supplier No. HY1003510MM 1ML
TTA-P2 is a selective, orally active, and brain-penetrant T-type calcium channel blocker used in neuroscience research to investigate neuronal excitability, pain mechanisms, and absence epilepsy. It is supplied as a 10 mM solution in DMSO or as solid material for assay preparation and in vivo studies.
  • Selective T-type calcium channel inhibitor with low-nanomolar potency (IC50 ≈ 22 nM).
  • Brain-penetrant profile suitable for in vivo studies.
  • Available as a 10 mM solution in DMSO and as solid quantities for flexible dosing.
  • High purity appropriate for research applications.
  • Used in studies of pain, tremor, and epilepsy mechanisms.