Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichloronitrobenzene, MP Biomedicals

Catalog No. ICN210132 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210132 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210132 Supplier MP Biomedicals, Inc Supplier No. 0521013210

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 618-62-2
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 191.995
InChI Key RNABGKOKSBUFHW-UHFFFAOYSA-N
Synonym 3,5-dichloronitrobenzene, benzene, 1,3-dichloro-5-nitro, m-dichloronitrobenzene, meta-dichloronitrobenzene, 3,5-dichloro-1-nitrobenzene, benzene,3-dichloro-5-nitro, pubchem3695, acmc-1b7uq, 3,5-dichloro-nitro-benzene, ksc493o5p
PubChem CID 12064
IUPAC Name 1,3-dichloro-5-nitrobenzene
SMILES C1=C(C=C(C=C1Cl)Cl)[N+](=O)[O-]

Specifications

Quantity 10 g
Formula Weight 192
Chemical Name or Material 3, 5-Dichloronitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.