Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3,5-Dichlorophenyl)(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate 95.0+%, TCI America™
SDP

Catalog No. p-7128674 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5145200MG 200 mg
D51451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5145200MG Supplier TCI America Supplier No. D5145200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1868173-25-4
Molecular Formula C22H21Cl2IO6S
Molecular Weight (g/mol) 611.268
InChI Key AYHVNGRMSLNAJQ-UHFFFAOYSA-M
Synonym (3,5-Dichlorophenyl)(2,4,6-trimethoxyphenyl)iodonium Tosylate
PubChem CID 131697914
IUPAC Name (3,5-dichlorophenyl)-(2,4,6-trimethoxyphenyl)iodanium;4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)[O-].COC1=CC(=C(C(=C1)OC)[I+]C2=CC(=CC(=C2)Cl)Cl)OC

Specifications

Melting Point 194°C
Quantity 200 mg
Formula Weight 611.27
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (3,5-Dichlorophenyl)(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.