Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichlorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D29095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29095G Supplier TCI America Supplier No. D29095G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67492-50-6
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.814
MDL Number MFCD00051935
InChI Key DKYRKAIKWFHQHM-UHFFFAOYSA-N
Synonym 3,5-dichlorophenyl boronic acid, 3,5-dichlorobenzeneboronic acid, 3,5-dichlorophenyl boranediol, 3,5-dichlophenylboronic acid, 3,5-dichloro phenylboronic acid, 3,5-dichlorobenzene boronic acid, 3,5-dichlorophenyl dihydroxyborane, boronic acid, b-3,5-dichlorophenyl, 3,5-dichlorophenylboronicacid
PubChem CID 2734331
IUPAC Name (3,5-dichlorophenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)Cl)Cl)(O)O

Specifications

Melting Point 306°C
Color White
Quantity 5 g
Formula Weight 190.81
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichlorophenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.