Learn More
Sigma Aldrich 3,5-Dichlorophenyldiazonium tetrafluoroborate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Reactant for:
- Electron transfer chemistry
- Microwave-accelerated cross-coupling reactions
- Sandmeyer bromination for preparation of bromobenzene derivatives
- Catalytic thiocyanation in the presence of copper salts
- Electrochemical reduction
Specifications
Specifications
| CAS | 350-67-4 |
| Molecular Formula | C6H3Cl2N2 -+ BF4 |
| Linear Formula | C6H3Cl2N2 -+ BF4 |
| MDL Number | MFCD00063489 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 260.81 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.