Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichloropyridine 98.0+%, TCI America™
SDP

Catalog No. D041125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D041125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D041125G Supplier TCI America Supplier No. D041125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2457-47-8
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00006376
InChI Key WPGHPGAUFIJVJF-UHFFFAOYSA-N
Synonym pyridine, 3,5-dichloro, ccris 1719, 3.5-dichloropyridine, 3,5-dichloro pyridine, 3,5-dichloro-pyridine, pubchem13506, acmc-1cqt0, wln: t6nj cg eg, 5-20-05-00417 beilstein handbook reference, ksc493c0d
PubChem CID 17153
IUPAC Name 3,5-dichloropyridine
SMILES ClC1=CC(Cl)=CN=C1

Specifications

Melting Point 65°C
Color White-Yellow
Boiling Point 178°C
Quantity 25 g
Formula Weight 147.99
Percent Purity ≥98.0% (GC)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichloropyridine
RTECSNumber US8575000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.