Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichloropyridine, 98%

Catalog No. AAL0338014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0338014 25 g
AAL0338022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0338014 Supplier Thermo Scientific Chemicals Supplier No. L0338014
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2457-47-8 | C5H3Cl2N | 147.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2457-47-8
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00006376
InChI Key WPGHPGAUFIJVJF-UHFFFAOYSA-N
Synonym pyridine, 3,5-dichloro, ccris 1719, 3.5-dichloropyridine, 3,5-dichloro pyridine, 3,5-dichloro-pyridine, pubchem13506, acmc-1cqt0, wln: t6nj cg eg, 5-20-05-00417 beilstein handbook reference, ksc493c0d
PubChem CID 17153
IUPAC Name 3,5-dichloropyridine
SMILES ClC1=CC(Cl)=CN=C1

Specifications

Melting Point 63°C to 67°C
Density 1.39
Boiling Point 178°C
Flash Point 74°C (165°F)
Odor Unpleasant
Quantity 25 g
UN Number UN2811
Beilstein 1973
Formula Weight 147.99
Percent Purity 98%
Chemical Name or Material 3,5-Dichloropyridine
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 219-537-9
RTECSNumber US8575000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.