Learn More
AdipoGen 3 5-Dichlorosalicylaldehyde
SDP
Supplier:  AdipoGen CDXD1064G025
Encompass_Preferred
Chemical. CAS 90-60-8. Formula C7H4Cl2O2. MW 191.01. 3,5-Dichlorosalicylaldehyde has been used as a building block or a reactant in the preparation of Schiff base ligands and RS-3,5-dichloro-2-1-phenylethyliminomethylphenol. 3,5-Dichlorosalicylaldehyde is used as a Mg2 fluorescent sensor. It works based on the Mg2-induced formation of a 21 3,5-Dichlorosalicylaldehyde-Mg2 complex, producing a chelation-enhanced fluorescence effect.
Catalog No. 50-256-1015