Learn More
Medchemexpress LLC 3,5-diethoxycarbonyl-1,4-dihydro-2,4,6-collidine | 632-93-9 | MFCD00005950 | >98.0% | 267.33 g·mol⁻¹ | C14H21NO4 | 1 G
SDP
Catalog No. 5000229095
Encompass_Preferred
Diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate (CAS 632-93-9) is a diethoxycarbonyl-substituted dihydropyridine used as a research reagent that interferes with heme synthesis and is employed in studies of porphyrin metabolism and ferrochelatase activity.
  • High purity: >98.0% (GC).
  • Chemical formula C14H21NO4; molecular weight 267.33 g·mol⁻¹.
  • Available in gram-scale quantities (1 G, 5 G, 25 G).
  • Used as a biochemical tool to block heme synthesis in cellular and in vivo studies.
  • Solid with reported melting point 130-132 °C for handling and storage considerations.
Catalog No. 50-002-29095 Supplier Medchemexpress LLC Supplier No. HY1185941G
Diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate (CAS 632-93-9) is a diethoxycarbonyl-substituted dihydropyridine used as a research reagent that interferes with heme synthesis and is employed in studies of porphyrin metabolism and ferrochelatase activity.
  • High purity: >98.0% (GC).
  • Chemical formula C14H21NO4; molecular weight 267.33 g·mol⁻¹.
  • Available in gram-scale quantities (1 G, 5 G, 25 G).
  • Used as a biochemical tool to block heme synthesis in cellular and in vivo studies.
  • Solid with reported melting point 130-132 °C for handling and storage considerations.