Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,5-diethyl-1,2-dihydro-1-phenyl-2-propylpyridine

Catalog No. CDS013696 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS013696 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS013696 Supplier Sigma Aldrich Supplier No. CDS013696500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,n-diisopropylaniline is an n,n-dialkylaniline
  • Its borane adducts have been prepared and reported as hydroborating agents
  • Thermal dissociation of the borane-n,n-diisopropylaniline adduct has been reported to afford gaseous diborane
  • N,n-diisopropylaniline is a diisoproplyamino derivative that can be prepared by reacting bromobenzene with diisopropylamine.n,n-diisopropylaniline was used for the synthesis of 4-diisopropylamino benzonitrile
  • It may be used for the synthesis of 6-(4-bromophenyl)-3-methoxy-5-methyl-8-oxabicyclo[3.2.1]octa-3,6-dien-2-one and n,n,nα,nα-tetraisopropylbenzidine

Specifications

CAS 34562-31-7
Molecular Formula C18H25N
Linear Formula C18H25N
MDL Number MFCD00071789
Quantity 1 g
Molecular Weight (g/mol) 255.4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.