Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Difluorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog No. AC431000050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC431000050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431000050 Supplier Thermo Scientific Chemicals Supplier No. 431000050

missing translation for 'validMainframeMsg'

CAS: 210532-25-5 | Molecular Formula: C6H3ClF2O2S | Molecular Weight: 212.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210532-25-5
Molecular Formula C6H3ClF2O2S
Molecular Weight (g/mol) 212.59
MDL Number MFCD02091378
InChI Key IIQKIICAOXAXEJ-UHFFFAOYSA-N
Synonym 3,5-difluorobenzenesulphonyl chloride, 3,5-difluorobenzene-1-sulfonyl chloride, 3,5-difluorobenzenesulfonylchloride, 3,5-difluorobenzenesulphonylchloride, benzenesulfonyl chloride, 3,5-difluoro, 3,5-difluoro-benzenesulfonyl chloride, pubchem8463, acmc-209fh1, ksc201g8f, buttpark 27\07-03
PubChem CID 2774058
IUPAC Name 3,5-difluorobenzenesulfonyl chloride
SMILES FC1=CC(=CC(F)=C1)S(Cl)(=O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 55.9°C to 58.2°C
Color Orange to Tan
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 212.6
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Difluorobenzenesulfonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.