Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Difluorobenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. D255910G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D255910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D255910G Supplier TCI America Supplier No. D255910G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79538-20-8
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.121
MDL Number MFCD00010313
InChI Key LOGIHEKXJKHXEC-UHFFFAOYSA-N
Synonym 3,5-difluorobenzyl alcohol, 3,5-difluorophenyl methanol, benzenemethanol, 3,5-difluoro, 3,5-difluorophenyl methan-1-ol, 3,5-difluorobenzylalcohol, 3, 5-difluorobenzyl alcohol, pubchem4918, acmc-1baxh, ksc493g5j, 3,5-difluoro-phenyl methanol
PubChem CID 522721
IUPAC Name (3,5-difluorophenyl)methanol
SMILES C1=C(C=C(C=C1F)F)CO

Specifications

Quantity 10 g
Formula Weight 144.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Difluorobenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.