Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Difluorobenzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. D26315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26315G 5 g
D263125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26315G Supplier TCI America Supplier No. D26315G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 141776-91-2
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.02
MDL Number MFCD00010304
InChI Key KVSVNRFSKRFPIL-UHFFFAOYSA-N
Synonym 3,5-difluorobenzyl bromide, 1-bromomethyl-3,5-difluorobenzene, 3,5-difluorobenzylbromide, alpha-bromo-3,5-difluorotoluene, 3,5-difluoro benzyl bromide, 3,5-bifluorobenzyl bromide, 1-bromomethyl-3,5-difluoro-benzene, benzene, 1-bromomethyl-3,5-difluoro, 5-bromomethyl-1,3-difluorobenzene, pubchem2245
PubChem CID 518896
IUPAC Name 1-(bromomethyl)-3,5-difluorobenzene
SMILES FC1=CC(CBr)=CC(F)=C1

Specifications

Color Yellow
Boiling Point 65°C
Quantity 5 g
UN Number 3265
Formula Weight 207.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Difluorobenzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.