Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF

Catalog No. AAH54427AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54427AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH54427AE Supplier Thermo Scientific Chemicals Supplier No. H54427AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1417985-27-3 | C7H5BrF2Mg | 231.323 g/mol

It is a green alternative to Tetrahydrofuran. is capable of producing high concentrations of bromo, benzyl and allyl Grignard reagents. 2-MeTHF quickly produces clean water phase splits without forming emulsions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a green alternative to Tetrahydrofuran. is capable of producing high concentrations of bromo, benzyl and allyl Grignard reagents. 2-MeTHF quickly produces clean water phase splits without forming emulsions.

Solubilit
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1417985-27-3
Molecular Formula C7H5BrF2Mg
Molecular Weight (g/mol) 231.323
MDL Number MFCD11113509
InChI Key QAXANYLHEOPEGM-UHFFFAOYSA-M
Synonym 3,5-difluorobenzylmagnesium bromide, 0.25m in 2-methf, qpuypkpzikvhdr-uhfffaoysa-m, 3,5-difluorobenzylmagnesium bromide, 3,5-difluorobenzyl magnesium bromide, grignard reagent
PubChem CID 73995805
SMILES [CH2-]C1=CC(=CC(=C1)F)F.[Mg+2].[Br-]

Specifications

Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 231.32
Concentration or Composition (by Analyte or Components) 0.25M in 2-MeTHF
Physical Form Liquid
Chemical Name or Material 3,5-Difluorobenzylmagnesium bromide
Hazard Category H225-H302-H314-H335
Hazard Statement GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.