Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,5-Difluorophenylmagnesium Bromide Solution

Catalog No. 56102950ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-102-950ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 56-102-950ML Supplier Sigma Aldrich Supplier No. 56102950ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,5-Difluorophenylmagnesium bromide can be used:As a starting material in the synthesis of cis-2-phenyl-1-amino-cyclopentanes as AMPA receptor potentiators.To prepare copper(I) complexes, which are used as luminescent guest molecules in organic light-emitting diodes (OLEDs).As an intermediate in the synthesis of 2-aminoethylbenzofuran H3 receptor antagonists.Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 156006-28-9
Density 0.975 g/mL at 25°C
Boiling Point 65°C
Molecular Formula C6H2BrF3Mg
Linear Formula F3C6H2MgBr
MDL Number MFCD01311471
Quantity 50 mL
Molecular Weight (g/mol) 235.28 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.