Learn More
Medchemexpress LLC 3,5-dihydro-5-methyl-N-3-pyridinylbenzo[1,2-b:4,5-b']dipyrrole-1(2H)-carboxamide hydrochloride | 1197334-04-5 | 99.4% | 328.80 g/mol | C17H17ClN4O | 10 MG
SDP
Catalog No. 5000245959
Encompass_Preferred
SB 206553 hydrochloride is the hydrochloride salt of a selective 5-HT2B/5-HT2C serotonin receptor antagonist used in pharmacological research. It is supplied as a high-purity solid for in vitro and in vivo studies and is typically provided in small milligram pack sizes.
  • Selective 5-HT2B and 5-HT2C receptor antagonist.
  • High purity, 99.4%.
  • Hydrochloride salt form for improved solubility.
  • Supplied in small pack sizes suitable for research (e.g., 5 MG, 10 MG, 25 MG).
  • Recommended storage: sealed, away from moisture; solid at -20°C; in solvent, -80°C for long-term.
Catalog No. 50-002-45959 Supplier Medchemexpress LLC Supplier No. HY10313510MG
SB 206553 hydrochloride is the hydrochloride salt of a selective 5-HT2B/5-HT2C serotonin receptor antagonist used in pharmacological research. It is supplied as a high-purity solid for in vitro and in vivo studies and is typically provided in small milligram pack sizes.
  • Selective 5-HT2B and 5-HT2C receptor antagonist.
  • High purity, 99.4%.
  • Hydrochloride salt form for improved solubility.
  • Supplied in small pack sizes suitable for research (e.g., 5 MG, 10 MG, 25 MG).
  • Recommended storage: sealed, away from moisture; solid at -20°C; in solvent, -80°C for long-term.