Learn More
Cayman Chemical 3,5-Dihydroxybenzaldehyde, Purity: ≥98%, CAS: 26153-38-8, 1 g
SDP
Catalog No. 502978899
Encompass_Preferred
A building block; has been used in the synthesis of 2,4-dimethylbenzoylhydrazones with antileishmanial and antioxidant activities
Catalog No. 50-297-8899 Supplier Cayman Chemical Supplier No. 304451 G
A building block; has been used in the synthesis of 2,4-dimethylbenzoylhydrazones with antileishmanial and antioxidant activities