Learn More
Selleck Chemical LLC 3 5-Dihydroxybenzoic acid
SDP
Catalog No. 5000003801
Encompass_Preferred
3 5-Dihydroxybenzoic acid ( -resorcylic acid) is a primary metabolite of alkylresinols which has been hydrolyzed by liver enzymes during phase I metabolism after several cycles of beta oxidation
Catalog No. 50-000-03801 Supplier Selleck Chemical LLC Supplier No. S616025MG
3 5-Dihydroxybenzoic acid ( -resorcylic acid) is a primary metabolite of alkylresinols which has been hydrolyzed by liver enzymes during phase I metabolism after several cycles of beta oxidation