Learn More
Medchemexpress LLC 3,5-dihydroxybenzoic acid | 99-10-5 | MFCD00002512 | 154.12 g/mol | C7H6O4 | 250 MG
SDP
Catalog No. 5000302841
Encompass_Preferred
3,5-Dihydroxybenzoic acid is a small-molecule analytical standard used in research and analytical and biochemical applications. It is reported as a potential biomarker for food-consumption studies, acts as an agonist of hydroxycarboxylic acid receptor 1 (HCA1), and is a competitive inhibitor of tyrosine phenol-lyase (TPL). Intended for laboratory research use only.
  • Analytical standard suitable for research and analytical applications.
  • Reported agonist activity at hydroxycarboxylic acid receptor 1 (HCA1).
  • Competitive inhibitor of tyrosine phenol-lyase useful in biochemical assays.
  • Used as a biomarker in food-consumption and metabolic studies.
  • Available in small pack sizes for analytical workflows.
Catalog No. 50-003-02841 Supplier Medchemexpress LLC Supplier No. 250MGHYW015560R
3,5-Dihydroxybenzoic acid is a small-molecule analytical standard used in research and analytical and biochemical applications. It is reported as a potential biomarker for food-consumption studies, acts as an agonist of hydroxycarboxylic acid receptor 1 (HCA1), and is a competitive inhibitor of tyrosine phenol-lyase (TPL). Intended for laboratory research use only.
  • Analytical standard suitable for research and analytical applications.
  • Reported agonist activity at hydroxycarboxylic acid receptor 1 (HCA1).
  • Competitive inhibitor of tyrosine phenol-lyase useful in biochemical assays.
  • Used as a biomarker in food-consumption and metabolic studies.
  • Available in small pack sizes for analytical workflows.