Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dihydroxybenzoic Acid, TCI America™
SDP

Catalog No. D2554500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2554500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2554500G Supplier TCI America Supplier No. D2554500G

Specifications

CAS 99-10-5
Molecular Formula C7H6O4
MDL Number MFCD00002512
Quantity 500 g
Synonym alpha-resorcylic acid, 5-carboxyresorcinol, benzoic acid, 3,5-dihydroxy, unii-2wc5lmo6l1, 3,5-dihydroxy benzoic acid, 3,5-dihydroxy-benzoic acid, 3,5-dhba, 3,5-dihydroxybenzoicacid, a-resorcylic acid, .alpha.-resorcylic acid
InChI Key UYEMGAFJOZZIFP-UHFFFAOYSA-N
SMILES OC(=O)C1=CC(O)=CC(O)=C1
IUPAC Name 3,5-dihydroxybenzoic acid
Molecular Weight (g/mol) 154.12
PubChem CID 7424
ChEBI CHEBI:39912
Formula Weight 154.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dihydroxybenzoic Acid
Show More Show Less
EINECSNumber (3)-1616
RTECSNumber VH3708000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.