Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dihydroxybenzonitrile, MP Biomedicals

Catalog No. ICN226911 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN226911 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN226911 Supplier MP Biomedicals, Inc Supplier No. 0522691101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3147-62-4
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
InChI Key PLYYMFBDRBSPJZ-UHFFFAOYSA-N
Synonym benzamide, 3,5-dihydroxy, 3,5-dihydroxy-benzamide, alpha-resorcylamide, 3,5-resorcylic acidamide, 3,5-dihydroxybenzoic acid amide, .alpha.-resorcylamide, pubchem13928, a-resorcyl amide, acmc-1coua, ksc494k8l
PubChem CID 76604
IUPAC Name 3,5-dihydroxybenzamide
SMILES C1=C(C=C(C=C1O)O)C(=O)N

Specifications

Quantity 10 g
Formula Weight 135
Chemical Name or Material 3, 5-Dihydroxybenzonitrile
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.