Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dihydroxybenzyl Alcohol 97.0+%, TCI America™
SDP

Catalog No. D26565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26565G 5 g
D265625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26565G Supplier TCI America Supplier No. D26565G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29654-55-5
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00016867
InChI Key NGYYFWGABVVEPL-UHFFFAOYSA-N
Synonym 3,5-dihydroxybenzyl alcohol, 5-hydroxymethyl-1,3-benzenediol, 5-hydroxymethyl benzene-1,3-diol, 5-hydroxymethyl resorcinol, benzyl alcohol, 3,5-dihydroxy, 1,3-benzenediol, 5-hydroxymethyl, 3,5-dihydroxybenzylalcohol, 3,5-dihydroxy benzyl alcohol, pubchem7418, acmc-1cjog
PubChem CID 34661
IUPAC Name 5-(hydroxymethyl)benzene-1,3-diol
SMILES C1=C(C=C(C=C1O)O)CO

Specifications

Melting Point 186°C
Quantity 5 g
Formula Weight 140.14
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dihydroxybenzyl Alcohol
RTECSNumber DO1400000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.