Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diiodo-4-pyridone-1-acetic Acid, TCI America™
SDP

Catalog No. D176325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D176325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D176325G Supplier TCI America Supplier No. D176325G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 101-29-1
Molecular Formula C7H5I2NO3
Molecular Weight (g/mol) 404.93
MDL Number MFCD00006185
InChI Key PVBALTLWZVEAIO-UHFFFAOYSA-N
Synonym pelvirinic acid, 3,5-diiodo-4-pyridone-1-acetic acid, umbradilic acid, diodone acid, unii-nlx9tz649p, 1 4h-pyridineacetic acid, 3,5-diiodo-4-oxo, nlx9tz649p, 3,5-diiodo-4-oxopyridin-1 4h-yl acetic acid, 2-3,5-diiodo-4-oxopyridin-1 4h-yl acetic acid, 3,5-diiodo-4-pyridone-n-acetic acid
PubChem CID 9304
IUPAC Name 2-(3,5-diiodo-4-oxopyridin-1-yl)acetic acid
SMILES C1=C(C(=O)C(=CN1CC(=O)O)I)I

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 404.93
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diiodo-4-pyridone-1-acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.