Learn More
Cayman Chemical 3,5-Diiodo-L-thyronine, Purity: ≥98%, CAS: 1041-01-6, 1 g
SDP
Catalog No. 502910370
Encompass_Preferred
A thyroid hormone metabolite; binds to TRα1, TRβ1, and TRβ2 (EC50s = 0.8, 0.79, and 0.1 µM, respectively, in cell-free assays); stimulates oxygen consumption in perfused livers isolated from rats with sodium iodide-induced hypothyroidism; reduces subcutaneous and epididymal fat mass, as well as decreases serum leptin and cholesterol levels, in high-fat diet-induced obese mice at 2.5 µg/g per day
Catalog No. 50-291-0370 Supplier Cayman Chemical Supplier No. 352121 G
A thyroid hormone metabolite; binds to TRα1, TRβ1, and TRβ2 (EC50s = 0.8, 0.79, and 0.1 µM, respectively, in cell-free assays); stimulates oxygen consumption in perfused livers isolated from rats with sodium iodide-induced hypothyroidism; reduces subcutaneous and epididymal fat mass, as well as decreases serum leptin and cholesterol levels, in high-fat diet-induced obese mice at 2.5 µg/g per day