Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diiodo-L-tyrosine dihydrate, 99%, Thermo Scientific™

Catalog No. AC225610050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC225610050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC225610050 Supplier Thermo Scientific Chemicals Supplier No. 225610050

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18835-59-1
Molecular Formula C9H13I2NO5
Molecular Weight (g/mol) 469.01
MDL Number MFCD00150275
InChI Key YWAGQOOMOOUEGY-QVGJJETENA-N
Synonym 3,5-diiodo-l-tyrosine dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, 3,5-diiodotyrosine dihydrate, 3,5-dijodtyrosindihydrat, c9h9i2no3.2h2o, 3,5-diiodo-l-tyrosine hydrate, l-tyrosine,3,5-diiodo-, hydrate 1:2, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoicaciddihydrate, 2s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl-propanoic acid dihydrate
PubChem CID 2724330
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid;dihydrate
SMILES O.O.N[C@@H](CC1=CC(I)=C(O)C(I)=C1)C(O)=O

Specifications

Melting Point 200.0°C
Infrared Spectrum Authentic
Linear Formula HOC6H2I2CH2CH(NH2)COOH·2H2O
Quantity 5 g
Beilstein 14,IV,237
Specific Rotation Condition −20.70 (20.00°C c=2.5, 0.1N NaOH/H2O)
Specific Rotation -20.7
Formula Weight 469.01
Percent Purity 99%
Chemical Name or Material 3,5-Diiodo-L-tyrosine dihydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.