Learn More
Medchemexpress LLC HY-126236 5mg Medchemexpress, 3,5-Diiodothyropropionic acid CAS:1158-10-7 Purity:>98%
SDP
Catalog No. 501932615
Encompass_Preferred
Medchemexpress, HY-126236 5mg 3,5-Diiodothyropropionic acid CAS:1158-10-7 3,5-Diiodothyropropionic acid is a thyroid hormone analog, induces α-myosin heavy chain mRNA expression, binds to thyroid hormone receptor (TR) , with K a of 2.40 and 4.06 M -1 for TRα1 and TRβ1, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2615 Supplier Medchemexpress LLC Supplier No. HY1262365MG
Medchemexpress, HY-126236 5mg 3,5-Diiodothyropropionic acid CAS:1158-10-7 3,5-Diiodothyropropionic acid is a thyroid hormone analog, induces α-myosin heavy chain mRNA expression, binds to thyroid hormone receptor (TR) , with K a of 2.40 and 4.06 M -1 for TRα1 and TRβ1, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.