Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethoxy-4-hydroxycinnamic Acid 98.0+%, TCI America™
SDP

Catalog No. D176525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D176525G 25 g
D17655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D176525G Supplier TCI America Supplier No. D176525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 530-59-6
Molecular Formula C11H12O5
Molecular Weight (g/mol) 224.212
MDL Number MFCD00004401
InChI Key PCMORTLOPMLEFB-ONEGZZNKSA-N
Synonym sinapic acid, sinapinic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, sinapinate, sinapate, trans-sinapic acid, 4-hydroxy-3,5-dimethoxycinnamic acid, synapoic acid, 2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid, unii-p0i60993ec
PubChem CID 637775
ChEBI CHEBI:15714
IUPAC Name (E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=CC(=CC(=C1O)OC)C=CC(=O)O

Specifications

Melting Point 198°C
Quantity 25 g
Formula Weight 224.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethoxy-4-hydroxycinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.