Learn More
Medchemexpress LLC 3,5-dimethoxy-benzoic acid, 2-[(1-naphthalenylamino)carbonyl]hydrazide | 444932-31-4 | MFCD02372953 | 99.8% | 365.38 g·mol^-1 | C20H19N3O4 | 100 MG
SDP
Catalog No. 5000245611
Encompass_Preferred
TC-O 9311 is a small-molecule agonist of the orphan G protein-coupled receptor GPR139, reported to show potent receptor activity in cellular assays (EC50 ≈ 39 nM). It is supplied as a white to off-white solid with high purity for laboratory research use.
  • Potent GPR139 agonist (EC50 ≈ 39 nM).
  • Active in HEK293 calcium FLIPR assay (EC50 ≈ 43 nM).
  • High purity (99.8%).
  • White to off-white solid, with recommended refrigerated storage.
  • Molecular weight 365.38 g·mol^-1; formula C20H19N3O4.
  • Suitable for in vitro pharmacology and receptor profiling.
Catalog No. 50-002-45611 Supplier Medchemexpress LLC Supplier No. HY101777100MG
TC-O 9311 is a small-molecule agonist of the orphan G protein-coupled receptor GPR139, reported to show potent receptor activity in cellular assays (EC50 ≈ 39 nM). It is supplied as a white to off-white solid with high purity for laboratory research use.
  • Potent GPR139 agonist (EC50 ≈ 39 nM).
  • Active in HEK293 calcium FLIPR assay (EC50 ≈ 43 nM).
  • High purity (99.8%).
  • White to off-white solid, with recommended refrigerated storage.
  • Molecular weight 365.38 g·mol^-1; formula C20H19N3O4.
  • Suitable for in vitro pharmacology and receptor profiling.