Learn More
Accela Chembio Inc MF: C9H10O3 | MW: 166.17 | ?96% | BP: 151 celsius at 760 mmHg.
Catalog No. 50353226
Encompass_Preferred
MF: C9H10O3 | MW: 166.17 | ?96% | BP: 151 celsius at 760 mmHg.
Catalog No. 50-353-226 Supplier Accela Chembio Inc Supplier No. SY001676100G
MF: C9H10O3 | MW: 166.17 | ?96% | BP: 151 celsius at 760 mmHg.