Learn More
Medchemexpress LLC 3,5-Dimethoxybenzaldehyde | 7311-34-4 | 166.18 | 5 G
SDP
Catalog No. 5000448766
Encompass_Preferred
3,5-Dimethoxybenzaldehyde is an antifungal agent that exhibits activity against *Saccharomyces cerevisiae* cell wall integrity mutants (slt2Δ and bck1Δ) and *Aspergillus fumigatus* MAPK mutants (sakAΔ and mpkCΔ). This product should be stored at 4°C under nitrogen. When in solvent, it should be stored at -80°C for 6 months or -20°C for 1 month, also under nitrogen. For shipping, it can be at room temperature if the duration is less than 2 weeks.
  • Antifungal agent
  • Exhibits antifungal activity against *Saccharomyces cerevisiae* cell wall integrity mutants
  • Exhibits antifungal activity against *Aspergillus fumigatus* MAPK mutants
  • Causes higher sensitivity in *Aspergillus fumigatus* MAPK mutants with 100% growth inhibition compared to the wild type
  • Inhibits the growth of *Saccharomyces cerevisiae* cell integrity mutants, showing potent antifungal activity
Catalog No. 50-004-48766 Supplier Medchemexpress LLC Supplier No. HYW0076265G
3,5-Dimethoxybenzaldehyde is an antifungal agent that exhibits activity against *Saccharomyces cerevisiae* cell wall integrity mutants (slt2Δ and bck1Δ) and *Aspergillus fumigatus* MAPK mutants (sakAΔ and mpkCΔ). This product should be stored at 4°C under nitrogen. When in solvent, it should be stored at -80°C for 6 months or -20°C for 1 month, also under nitrogen. For shipping, it can be at room temperature if the duration is less than 2 weeks.
  • Antifungal agent
  • Exhibits antifungal activity against *Saccharomyces cerevisiae* cell wall integrity mutants
  • Exhibits antifungal activity against *Aspergillus fumigatus* MAPK mutants
  • Causes higher sensitivity in *Aspergillus fumigatus* MAPK mutants with 100% growth inhibition compared to the wild type
  • Inhibits the growth of *Saccharomyces cerevisiae* cell integrity mutants, showing potent antifungal activity