Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethoxybenzamide 98.0+%, TCI America™
SDP

Catalog No. D15585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D15585G Supplier TCI America Supplier No. D15585G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17213-58-0
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00007987
InChI Key YTLRWVNYANKXOW-UHFFFAOYSA-N
Synonym benzamide, 3,5-dimethoxy, acmc-1c3k0, 3,5-dimethoxybenzamide
PubChem CID 72878
IUPAC Name 3,5-dimethoxybenzamide
SMILES COC1=CC(=CC(OC)=C1)C(N)=O

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 5 g
Formula Weight 181.19
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethoxybenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.