Learn More
Medchemexpress LLC 3,5-dimethoxybenzoic acid 2-[(1-naphthalenylamino)carbonyl]hydrazide | 444932-31-4 | MFCD02372953 | ≥98.0% | 365.38 g·mol⁻¹ | C20H19N3O4 | 50 MG
SDP
Catalog No. 5000245685
Encompass_Preferred
TC-O 9311 is a potent, selective agonist of the orphan G protein-coupled receptor GPR139 used as a research tool in cell-based pharmacology. It was disclosed as Compound 1a in the primary literature and shows an EC50 of 39 nM in CHO-K1 cells expressing human GPR139. The compound is characterized for in vitro use and is typically supplied at high purity for pharmacological studies.
  • Potent GPR139 agonist (EC50 = 39 nM in CHO-K1 cells).
  • High selectivity with no cross-reactivity across a diverse 90-target panel.
  • Suitable as a positive control in cell-based pharmacology assays.
  • Supplied at high purity (≥98% by HPLC).
  • Well characterized: CAS 444932-31-4; MFCD02372953; formula C20H19N3O4; MW 365.38 g·mol⁻¹.
Catalog No. 50-002-45685 Supplier Medchemexpress LLC Supplier No. HY10177750MG
TC-O 9311 is a potent, selective agonist of the orphan G protein-coupled receptor GPR139 used as a research tool in cell-based pharmacology. It was disclosed as Compound 1a in the primary literature and shows an EC50 of 39 nM in CHO-K1 cells expressing human GPR139. The compound is characterized for in vitro use and is typically supplied at high purity for pharmacological studies.
  • Potent GPR139 agonist (EC50 = 39 nM in CHO-K1 cells).
  • High selectivity with no cross-reactivity across a diverse 90-target panel.
  • Suitable as a positive control in cell-based pharmacology assays.
  • Supplied at high purity (≥98% by HPLC).
  • Well characterized: CAS 444932-31-4; MFCD02372953; formula C20H19N3O4; MW 365.38 g·mol⁻¹.