Learn More
Medchemexpress LLC 3,5-dimethoxybenzyl alcohol | 705-76-0 | MFCD00004641 | 99.9% | 168.19 g/mol | C9H12O3 | 50 G
SDP
Catalog No. 5000299490
Encompass_Preferred
3,5-Dimethoxybenzyl alcohol is a substituted benzyl alcohol used as a research reagent and intermediate in synthetic organic chemistry. It is provided as a white to off-white solid, with CAS 705-76-0, molecular formula C9H12O3, and molecular weight 168.19 g/mol. Typical uses include organic synthesis and natural product research.
  • High purity: 99.90% (HPLC).
  • White to off-white solid appearance.
  • Identity verified by 1H NMR.
  • Stable when stored under recommended conditions.
  • Useful intermediate for organic synthesis and research applications.
Catalog No. 50-002-99490 Supplier Medchemexpress LLC Supplier No. HYW01709350G
3,5-Dimethoxybenzyl alcohol is a substituted benzyl alcohol used as a research reagent and intermediate in synthetic organic chemistry. It is provided as a white to off-white solid, with CAS 705-76-0, molecular formula C9H12O3, and molecular weight 168.19 g/mol. Typical uses include organic synthesis and natural product research.
  • High purity: 99.90% (HPLC).
  • White to off-white solid appearance.
  • Identity verified by 1H NMR.
  • Stable when stored under recommended conditions.
  • Useful intermediate for organic synthesis and research applications.