Learn More
Apexbio Technology LLC 3-5-DIMETHOXYPHENOL-1G
SDP
Catalog No. 5000568443
Encompass_Preferred
3 5-Dimethoxyphenol (CAS No 500-99-2) is a compound of interest in the biomedical research field originating from phenolic derivatives Its mechanism of action involves modulation of oxidative stress pathways where it acts as an antioxidant by neutralizing free radicals Researchers have observed its in vitro activity in the nanomolar to low micromolar range highlighting its potency in cellular assays Typical applications include investigations using cell models such as fibroblasts and hepatocytes as well as in animal studies to explore its potential therapeutic effects This compound is also utilized in drug screening to assess antioxidant properties Experimental concentrations typically depend on the specific research design ensuring targeted efficacy without toxicity
Catalog No. 50-005-68443 Supplier Apexbio Technology LLC Supplier No. C70361000
3 5-Dimethoxyphenol (CAS No 500-99-2) is a compound of interest in the biomedical research field originating from phenolic derivatives Its mechanism of action involves modulation of oxidative stress pathways where it acts as an antioxidant by neutralizing free radicals Researchers have observed its in vitro activity in the nanomolar to low micromolar range highlighting its potency in cellular assays Typical applications include investigations using cell models such as fibroblasts and hepatocytes as well as in animal studies to explore its potential therapeutic effects This compound is also utilized in drug screening to assess antioxidant properties Experimental concentrations typically depend on the specific research design ensuring targeted efficacy without toxicity