Learn More
Medchemexpress LLC 3,5-Dimethoxyphenol | 500-99-2 | 154.16 | 1 G

Supplier: Medchemexpress LLC HYW0011171G
3,5-Dimethoxyphenol, also known as Phloroglucinol Dimethyl Ether, is an endogenous metabolite and a toxin metabolite. It has been found in humans who have consumed yew leaves. This product is strictly for research use and not for sale to patients.
- Alternative names: Phloroglucinol Dimethyl Ether
- Classification: Phenol, monophenol
- Purity: 99.84%
- Molecular weight: 154.16
- Formula: C8H10O3
- CAS No.: 500-99-2
- Appearance: Solid below 40°C, liquid above 43°C
- Initial source: Plants from the Schisandraceae family (Schisandra sphenanthera Rehd. et Wils) and Taxaceae family (Taxus chinensis (Pilger) Rehd.)
- Application: Used in research to study metabolic diseases
- SMILES: OC1=CC(OC)=CC(OC)=C1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.