Learn More
Medchemexpress LLC 3,5-Dimethoxyphenol | 500-99-2 | 154.16 | 10 G
SDP
Catalog No. 5000447682
Encompass_Preferred
3,5-Dimethoxyphenol is an endogenous and toxin metabolite found in humans after consuming yew leaves. It is a research-use-only chemical reagent, not intended for human use, and is available in solid or liquid form. The compound can be stored at -20°C for 3 years in pure form or at -80°C for 6 months when in solvent. Solubility in DMSO is 100 mg/mL.
  • Found in human consuming yew leaves
  • Available in solid or liquid form
  • Solubility in DMSO is 100 mg/mL
  • Pure form storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-47682 Supplier Medchemexpress LLC Supplier No. HYW00111710G
3,5-Dimethoxyphenol is an endogenous and toxin metabolite found in humans after consuming yew leaves. It is a research-use-only chemical reagent, not intended for human use, and is available in solid or liquid form. The compound can be stored at -20°C for 3 years in pure form or at -80°C for 6 months when in solvent. Solubility in DMSO is 100 mg/mL.
  • Found in human consuming yew leaves
  • Available in solid or liquid form
  • Solubility in DMSO is 100 mg/mL
  • Pure form storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month