Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,5-Dimethoxyphenol | 500-99-2 | 154.16 | 25 G
SDP

Catalog No. 5000449917
Encompass_Preferred

3,5-Dimethoxyphenol is a toxin metabolite found in humans consuming yew leaves. This product is for research use only and is not intended for medical or clinical applications.

  • Target: Endogenous metabolite
  • Pathway: Metabolic enzyme/protease
  • Storage: Pure form stored at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month.
  • Solubility (in vitro): DMSO at 100 mg/mL (648.68 mM) requiring ultrasonication.

Catalog No. 50-004-49917 Supplier Medchemexpress LLC Supplier No. 25GHYW001117
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,5-Dimethoxyphenol is a toxin metabolite found in humans consuming yew leaves. This product is for research use only and is not intended for medical or clinical applications.

  • Target: Endogenous metabolite
  • Pathway: Metabolic enzyme/protease
  • Storage: Pure form stored at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month.
  • Solubility (in vitro): DMSO at 100 mg/mL (648.68 mM) requiring ultrasonication.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.