Learn More
Medchemexpress LLC 3,5-Dimethoxyphenol (Phloroglucinol dimethyl ether) | 500-99-2 | 154.16 | 5 G
SDP
Catalog No. 5000447680
Encompass_Preferred
3,5-Dimethoxyphenol is a toxin metabolite, found in humans who have consumed yew leaves. This compound is also known as Phloroglucinol Dimethyl Ether.
  • Toxin metabolite
  • Endogenous metabolite
  • Functions as an inhibitor
  • Appears as a solid below 40°C, liquid above 43°C
  • Soluble in DMSO
Catalog No. 50-004-47680 Supplier Medchemexpress LLC Supplier No. HYW0011175G
3,5-Dimethoxyphenol is a toxin metabolite, found in humans who have consumed yew leaves. This compound is also known as Phloroglucinol Dimethyl Ether.
  • Toxin metabolite
  • Endogenous metabolite
  • Functions as an inhibitor
  • Appears as a solid below 40°C, liquid above 43°C
  • Soluble in DMSO