Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethyl-1-phenylpyrazole 98.0+%, TCI America™
SDP

Catalog No. D28345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28345G 5 g
D283425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28345G Supplier TCI America Supplier No. D28345G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1131-16-4
Molecular Formula C11H12N2
Molecular Weight (g/mol) 172.231
MDL Number MFCD00020728
InChI Key ULPMPUPEFBDQQA-UHFFFAOYSA-N
Synonym 3,5-dimethyl-1-phenyl-1h-pyrazole, 1h-pyrazole, 3,5-dimethyl-1-phenyl, dimethylphenylpyrazole, unii-0sb52bxx7m, 1h-pyrazole,3,5-dimethyl-1-phenyl, 0sb52bxx7m, 1,3-dimethyl-5-phenylpyrazole, phenyldimethylpyrazole, enamine_001158, pyrazole, dimethylphenyl
PubChem CID 70800
IUPAC Name 3,5-dimethyl-1-phenylpyrazole
SMILES CC1=CC(=NN1C2=CC=CC=C2)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 172.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Dimethyl-1-phenylpyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.