Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC01201CB
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC01201CB 250 mg
CC01201DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC01201CB Supplier Thermo Scientific Chemicals Supplier No. CC01201CB
Add to Cart

Chemical Identifiers

CAS 113808-86-9
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD00159642
InChI Key IOOWDXMXZBYKLR-UHFFFAOYSA-N
Synonym 3,5-dimethylpyrazole-4-carboxylic acid, 1h-pyrazole-4-carboxylic acid, 3,5-dimethyl, 1h-pyrazole-4-carboxylicacid, 3,5-dimethyl, 3,5-dimethyl-1h-pyrazole-4-carboxylicacid, acmc-20a7j9, pyrazole carboxylic acid 1, ksc173i7f, 3,5-dimethylpyrazole-4-carboxylic, 3,5-dimethylpyrazole-4-carboxylicacid, 1h-pyrazole-4-carboxylicacid,3,5-dimethyl
PubChem CID 2776164
IUPAC Name 3,5-dimethyl-1H-pyrazole-4-carboxylic acid
SMILES CC1=C(C(=NN1)C)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 3, 5-Dimethyl-1H-pyrazole-4-carboxylic acid
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.