Learn More
3,5-Dimethyl-1H-pyrazole, 99%
Description
3,5-Dimethyl-1H-pyrazole is combined with Chromium(VI) oxide, is a valuable reagent for oxidation of primary and secondary alcohols to carbonyl compounds. It reacts with malonic esters gives a family of cross-conjugated monomeric betaines. It is a common reagent for the preparation of pyrazolato ligated complexes. It is also used to prepare N-1-substituted derivatives having antibacterial activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3,5-Dimethyl-1H-pyrazole is combined with Chromium(VI) oxide, is a valuable reagent for oxidation of primary and secondary alcohols to carbonyl compounds. It reacts with malonic esters gives a family of cross-conjugated monomeric betaines. It is a common reagent for the preparation of pyrazolato ligated complexes. It is also used to prepare N-1-substituted derivatives having antibacterial activity.
Solubility
Soluble in water, methanol 0.1 g/mL.
Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.

Specifications
Specifications
Melting Point | 105°C to 108°C |
pH | 6.7 |
Boiling Point | 218°C |
Quantity | 250 g |
Beilstein | 106325 |
Solubility Information | Soluble in water,methanol 0.1g/mL. |
Formula Weight | 96.13 |
Percent Purity | 99% |
Chemical Name or Material | 3,5-Dimethyl-1H-pyrazole |
Safety and Handling
Hazard Category | H302-H361fd-H373 |
Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c |
EINECSNumber | 200-657-5 |
RTECSNumber | UQ6477600 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.