Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethyl-4-nitro-2-pyridinemethanol 98.0+%, TCI America™
SDP

Catalog No. D40265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40265G 5 g
D402625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40265G Supplier TCI America Supplier No. D40265G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 149082-03-1
Molecular Formula C8H10N2O3
Molecular Weight (g/mol) 182.179
MDL Number MFCD03426035
InChI Key KBCDOXSSYLFMHH-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-nitropyridin-2-yl methanol, 3,5-dimethyl-2-hydroxymethyl-4-nitropyridine, 3,5-dimethyl-4-nitropyridine-2-methanol, 3,5-dimethyl-4-nitro-2-pyridinemethanol, 2-hydroxymethyl-3,5-dimethyl-4-nitropyridine, 3,5-dimethyl-4-nitro-2-pyridyl-1-methanol, 2-pyridinemethanol, 3,5-dimethyl-4-nitro, 3,5-dimethyl-4-nitro-2-pyridyl methan-1-ol, 4-nitro-3,5-dimethyl-2-hydroxymethyl pyridine
PubChem CID 4049415
IUPAC Name (3,5-dimethyl-4-nitropyridin-2-yl)methanol
SMILES CC1=CN=C(C(=C1[N+](=O)[O-])C)CO

Specifications

Melting Point 68°C
Quantity 5 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethyl-4-nitro-2-pyridinemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.