Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethyl-4-propoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D50805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5080-5G 5 g
D50801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5080-5G Supplier TCI America Supplier No. D50805G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 357611-51-9 | C11H17BO3 | 208.064 g/mol

Specifications

Chemical Name or Material 3,5-Dimethyl-4-propoxyphenylboronic Acid (contains varying amounts of Anhydride)
CAS 357611-51-9
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Molecular Formula C11H17BO3
MDL Number MFCD05865178
Synonym 3,5-dimethyl-4-propoxyphenyl boronic acid, 4-propoxy-3,5-dimethylphenylboronic acid, boronicacid, b-3,5-dimethyl-4-propoxyphenyl, 3,5-dimethyl-4-n-propoxybenzeneboronic acid, 4-propoxy-3,5-dimethylbenzeneboronic acid
InChI Key ISUPZUFVQLUFLM-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C(=C1)C)OCCC)C)(O)O
IUPAC Name (3,5-dimethyl-4-propoxyphenyl)boronic acid
Molecular Weight (g/mol) 208.064
PubChem CID 16217512
Formula Weight 208.06
Percent Purity 97.0% to 110.0%
Grade Not Available
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.