Learn More
AdipoGen 3 5-Dimethyl-NAPQI
SDP
Supplier:  AdipoGen CDXA0163M010
Encompass_Preferred
Chemical. CAS 74827-85-3. Formula C10H11NO2. MW 177.2. Synthetic Metabolite. Oxidizing analog of N-acetyl-p-benzoquinone imine NAPQI, a toxic metabolite of acetaminophen paracetamol. Primarily oxidizes protein thiols. Shows cytotoxic properties, through disruption of intracellular Ca2 homeostasis. Compound can be used as analytical reference material.
Catalog No. 50-256-1822