Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',5'-Dimethylacetophenone 97.0+%, TCI America™
SDP

Catalog No. D48685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48685G 5 g
D48681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48685G Supplier TCI America Supplier No. D48685G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5379-16-8
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD01075693
InChI Key BKIHFZLJJUNKMZ-UHFFFAOYSA-N
Synonym 1-3,5-dimethylphenyl ethanone, 3,5-dimethylacetophenone, 3',5'-dimethylacetophenone, 1-3,5-dimethylphenyl ethan-1-one, ethanone, 1-dimethylphenyl, ethanone, 1-3,5-dimethylphenyl, 1-dimethylphenyl ethan-1-one, pubchem24402, 3', 5'-dimethylacetophenone, 3',5'-dimethyl acetophenone
PubChem CID 14921
IUPAC Name 1-(3,5-dimethylphenyl)ethanone
SMILES CC1=CC(=CC(=C1)C(=O)C)C

Specifications

Color White
Boiling Point 237°C
Quantity 5 g
Formula Weight 148.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3′,5′-Dimethylacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.