Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylanisole 98.0+%, TCI America™
SDP

Catalog No. D244125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D244125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D244125ML Supplier TCI America Supplier No. D244125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory reagent, Colorless to light yellow liquid with a purity of gt;98.0% by GC.

Chemical Identifiers

CAS 874-63-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00008398
InChI Key JCHJBEZBHANKGA-UHFFFAOYSA-N
Synonym 3,5-dimethylanisole, 5-methoxy-m-xylene, benzene, 1-methoxy-3,5-dimethyl, 1-methoxy-3,5-dimethyl-benzene, 3,5-dimethylanisol, 3,5-dimethylanizole, acmc-1bkxt, anisole, 3,5-dimethyl, ksc494e7h, 5-methoxy-1,3-dimethylbenzene
PubChem CID 70126
IUPAC Name 1-methoxy-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)OC)C

Specifications

Color Yellow
Quantity 25 mL
Physical Form Clear Liquid at 20°C
Boiling Point 193°C
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Grade Not Available
Chemical Name or Material 3,5-Dimethylanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.