Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylbenzoic Acid 99.0+%, TCI America™
SDP

Catalog No. D068425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D068425G 25 g
D0684250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D068425G Supplier TCI America Supplier No. D068425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 499-06-9 | C9H10O2 | 150.177 g/mol

Chemical Identifiers

CAS 499-06-9
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002525
InChI Key UMVOQQDNEYOJOK-UHFFFAOYSA-N
Synonym mesitylenic acid, benzoic acid, 3,5-dimethyl, unii-ed8av34n0y, 3,5-dimethyl benzoic acid, 3,5-dimethyl-benzoic acid, ed8av34n0y, 3,5-dimethylbenzoicacid, pubchem15440, 3,5-dimethylbezoic acid, 3,5 dimethylbenzoic acid
PubChem CID 10356
ChEBI CHEBI:64821
IUPAC Name 3,5-dimethylbenzoic acid
SMILES CC1=CC(=CC(=C1)C(=O)O)C

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 150.18
Percent Purity ≥99.0% (T)
Grade Not Available
Chemical Name or Material 3,5-Dimethylbenzoic Acid
EINECSNumber (3)-4260
RTECSNumber DG8734030
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.