Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylcyclohexanone (mixture of isomers) 98.0+%, TCI America™
SDP

Catalog No. D12825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12825ML 5 mL
D128225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D12825ML Supplier TCI America Supplier No. D12825ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2320-30-1
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00019467
InChI Key MSANHHHQJYQEOK-UHFFFAOYSA-N
PubChem CID 137544
IUPAC Name 3,5-dimethylcyclohexan-1-one
SMILES CC1CC(CC(=O)C1)C

Specifications

Color Yellow
Boiling Point 176°C
Quantity 5 mL
UN Number 1224
Formula Weight 126.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Dimethylcyclohexanone (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.