Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,5-dimethylisoxazole-4-boronic acid pinacol ester | 832114-00-8 | 99.07% | C11H18BNO3 | 1 ML
SDP

Catalog No. 5000434879
Encompass_Preferred

3,5-dimethylisoxazole-4-boronic acid pinacol ester is a drug intermediate. It is utilized in the synthesis of bromodomain inhibitors.

  • Drug intermediate
  • Used in the synthesis of bromodomain inhibitors
  • Powder stable for up to 3 years at -20°C or 2 years at 4°C
  • Solution stable for up to 2 years at -80°C or 1 year at -20°C
  • Appearance: Off-white to light yellow solid
  • Purity: 99.07% (HPLC)

Catalog No. 50-004-34879 Supplier Medchemexpress LLC Supplier No. HY7819710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,5-dimethylisoxazole-4-boronic acid pinacol ester is a drug intermediate. It is utilized in the synthesis of bromodomain inhibitors.

  • Drug intermediate
  • Used in the synthesis of bromodomain inhibitors
  • Powder stable for up to 3 years at -20°C or 2 years at 4°C
  • Solution stable for up to 2 years at -80°C or 1 year at -20°C
  • Appearance: Off-white to light yellow solid
  • Purity: 99.07% (HPLC)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.