Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,5-Dimethylisoxazole-4-boronic acid pinacol ester | 832114-00-8 | 99.1% | C11H18BNO3 | 1 G
SDP

Catalog No. 5000433246
Encompass_Preferred

3,5-Dimethylisoxazole-4-boronic acid pinacol ester is a drug intermediate used for the synthesis of bromodomain inhibitors. This product has been tested and complies with given specifications, though it is intended for research use only.

  • Drug intermediate for synthesis of bromodomain inhibitors
  • Purity of 99.1% (HPLC)
  • Appears as an off-white to light yellow solid
  • Chemical formula C11H18BNO3
  • Molecular weight 223.08
  • 1H NMR spectrum consistent with structure

Catalog No. 50-004-33246 Supplier Medchemexpress LLC Supplier No. HY781971G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,5-Dimethylisoxazole-4-boronic acid pinacol ester is a drug intermediate used for the synthesis of bromodomain inhibitors. This product has been tested and complies with given specifications, though it is intended for research use only.

  • Drug intermediate for synthesis of bromodomain inhibitors
  • Purity of 99.1% (HPLC)
  • Appears as an off-white to light yellow solid
  • Chemical formula C11H18BNO3
  • Molecular weight 223.08
  • 1H NMR spectrum consistent with structure

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.